Anti-GPR171 Rabbit Polyclonal Antibody

Proveedor: Bioss
BS-6564R
BOSSBS-6564REA 725 EUR
BOSSBS-6564R
Anti-GPR171 Rabbit Polyclonal Antibody
Anticuerpos
Platelet Activating Receptor H963 is an orphan receptor that is a member of the Platelet Activating Factor Receptor subfamily. ESTs have been isolated from normal human blood (activated), liver, and testis libraries, and from a human uterine adenocarcinoma library.

WB(1:100-1000), FCM(1:20-100), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: GPR171
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR