Anti-BCAT1 Rabbit Polyclonal Antibody (Cy5®)

Proveedor: Bioss
BS-6613R-CY5
BOSSBS-6613R-CY5EA 900 EUR
BOSSBS-6613R-CY5
Anti-BCAT1 Rabbit Polyclonal Antibody (Cy5®)
Anticuerpos
Catalyzes the first reaction in the catabolism of the essential branched chain amino acids leucine, isoleucine, and valine.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCAT1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR