Anti-WDR16 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Proveedor: Bioss
BS-6750R-A750
BOSSBS-6750R-A750EA 915 EUR
BOSSBS-6750R-A750
Anti-WDR16 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
WD repeat containing proteins, such as WDR16, play crucial roles in a wide range of physiologic functions, including signal transduction, RNA processing, remodeling the cytoskeleton, regulation of vesicular traffic, and cell division.

Type: Primary
Antigen: WDR16
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 401-500/620
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

ESPECIFICACIONES

Environmentally Preferable
Nombre del antígeno WDR16
Sinónimos de antígenos WDRPUH|WD40-repeat protein up-regulated in HCC|WD repeat-containing protein 16|WDR16
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 750
Reactividad Human
Huésped Rabbit
ID del gen 146845
Isotipo IgG
Western blot Yes
InmunoQuímica Yes
FluorescenciaInmunológica Yes
Modificación Unmodified
Epítopo 401-500/620
Inmunógeno KLH conjugated synthetic peptide derived from human WDR16
Forma Liquid
Molecular weight 68 kDa
Purification Purified by Protein A
Concentración 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Temperatura de almacenamiento Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR