Anti-CIDEC Rabbit Polyclonal Antibody

Proveedor: Bioss
BS-6796R
BOSSBS-6796REA 725 EUR
BOSSBS-6796R
Anti-CIDEC Rabbit Polyclonal Antibody
Anticuerpos
Type: Primary
Antigen: CIDEC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR