Anti-SDCCAG8 Rabbit Polyclonal Antibody (Cy3®)

Proveedor: Bioss
BS-7011R-CY3
BOSSBS-7011R-CY3EA 900 EUR
BOSSBS-7011R-CY3
Anti-SDCCAG8 Rabbit Polyclonal Antibody (Cy3®)
Anticuerpos
The exact function of SDCCAG8 remains unknown. It is expressed in thymus, prostate, testis, ovary, small intestine, colon, mucosa, colon and renal cancer tumors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SDCCAG8
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR