Anti-SDCCAG8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Proveedor: Bioss
BS-7011R-FITC
BOSSBS-7011R-FITCEA 900 EUR
BOSSBS-7011R-FITC
Anti-SDCCAG8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
The exact function of SDCCAG8 remains unknown. It is expressed in thymus, prostate, testis, ovary, small intestine, colon, mucosa, colon and renal cancer tumors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SDCCAG8
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR