Anti-SVIL Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Proveedor: Bioss
BS-7123R-A555
BOSSBS-7123R-A555EA 893 EUR
BOSSBS-7123R-A555
Anti-SVIL Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticuerpos
Forms a high-affinity link between the actin cytoskeleton and the membrane. Isoform 1 (archvillin) is among the first costameric proteins to assemble during myogenesis and it contributes to myogenic membrane structure and differentiation. Appears to be involved in myosin II assembly. May modulate myosin II regulation through MLCK during cell spreading, an initial step in cell migration. May play a role in invadopodial function. Isoform 2 may be involved in modulation of focal adhesions. Supervillin-mediated down-regulation of focal adhesions involves binding to TRIP6. Plays a role inMore Product Information