Anti-GGCX Rabbit Polyclonal Antibody
Proveedor: Bioss
BS-7759R
BOSSBS-7759REA 1090 EUR
BOSSBS-7759R
Anti-GGCX Rabbit Polyclonal Antibody
Anticuerpos
GGCX is a 758 amino acid multi-pass membrane protein. Localized to the membrane of the endoplasmic reticulum, GGCX functions to mediate the vitamin K-dependent carboxylation of glutamate residues on target proteins, thereby producing calcium binding gamma-carboxyglutamate (Gla) residues on these proteins and simultaneously converting vitamin K to vitamin K epoxide. GGCX exists as a monomer and, via its ability to modify glutamate residues, it accomplishes the post-translational changes that are necessary for the activity of all vitamin K-dependent proteins (such as blood coagulation and boneMore Product Information