Anti-UCHL5IP Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Proveedor: Bioss
BS-7776R-A488
BOSSBS-7776R-A488EA 893 EUR
BOSSBS-7776R-A488
Anti-UCHL5IP Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
Contributes to mitotic spindle assembly, maintenance of centrosome integrity and completion of cytokinesis as part of the HAUS augmin-like complex.Tissue specificity:Detected in spleen, thymus, testis, ovary, small intestine and colon, with highest levels of expression in testis and ovary.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: UCHL5IP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR