Anti-PELO Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Proveedor: Bioss
BS-7821R-A750
BOSSBS-7821R-A750EA 893 EUR
BOSSBS-7821R-A750
Anti-PELO Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
Required for normal chromosome segregation during cell division and genomic stability (By similarity). May function in recognising stalled ribosomes and triggering endonucleolytic cleavage of the mRNA, a mechanism to release non-functional ribosomes and degrade damaged mRNAs. May have ribonuclease activity.

Type: Primary
Antigen: PELO
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 141-240/385
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR