Anti-UBE2V1 Rabbit Polyclonal Antibody (Cy5®)
Proveedor: Bioss
BS-7882R-CY5
BOSSBS-7882R-CY5EA 900 EUR
BOSSBS-7882R-CY5
Anti-UBE2V1 Rabbit Polyclonal Antibody (Cy5®)
Anticuerpos
Has no ubiquitin ligase activity on its own. The UBE2V1-UBE2N heterodimer catalyzes the synthesis of non-canonical poly-ubiquitin chains that are linked through Lys-63. This type of poly-ubiquitination activates IKK and does not seem to involve protein degradation by the proteasome. Plays a role in the activation of NF-kappa-B mediated by IL1B, TNF, TRAF6 and TRAF2. Mediates transcriptional activation of target genes. Plays a role in the control of progress through the cell cycle and differentiation. Plays a role in the error-free DNA repair pathway and contributes to the survival of cellsMore Product Information