Anti-CMTM8 Rabbit Polyclonal Antibody (Cy5®)

Proveedor: Bioss
BS-8024R-CY5
BOSSBS-8024R-CY5EA 900 EUR
BOSSBS-8024R-CY5
Anti-CMTM8 Rabbit Polyclonal Antibody (Cy5®)
Anticuerpos
This gene belongs to the chemokine-like factor gene superfamily, a novel family that is similar to the chemokine and the transmembrane 4 superfamilies. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 3. This gene is widely expressed in many tissues, but the exact function of the encoded protein is unknown. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CMTM8
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR