Anti-FAM102B Rabbit Polyclonal Antibody (Cy7®)

Proveedor: Bioss
BS-8230R-CY7
BOSSBS-8230R-CY7EA 900 EUR
BOSSBS-8230R-CY7
Anti-FAM102B Rabbit Polyclonal Antibody (Cy7®)
Anticuerpos
Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. TheMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno FAM102B
Nombre del antígeno Family With Sequence Similarity 102 Member B
Environmentally Preferable
Sinónimos de antígenos SYM-3B
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Cy7®
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 284611
Isotipo IgG
InmunoQuímica Yes
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR