Anti-DEPTOR Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Proveedor: Bioss
BS-8255R-A750
BOSSBS-8255R-A750EA 915 EUR
BOSSBS-8255R-A750
Anti-DEPTOR Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anticuerpos
DEPTOR (DEP domain containing MTOR-interacting protein), also known as DEP.6 or DEPDC6 (DEP domain-containing protein 6), is a 409 amino acid protein that negatively regulates mTORC1 and mTORC2 pathways. DEPTOR interacts with FRAP via its PDZ domain, and undergoes post-translational phosphorylation. Containing two DEP domains and one PDZ (DHR) domain, DEPTOR is encoded by a gene that maps to human chromosome 8q24.12. Chromosome 8 consists of nearly 146 million base pairs, encodes over 800 genes and is associated with a variety of diseases and malignancies. Schizophrenia, bipolar disorder,More Product Information