Anti-DPY19L4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Proveedor: Bioss
BS-8292R-A488
BOSSBS-8292R-A488EA 893 EUR
BOSSBS-8292R-A488
Anti-DPY19L4 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
Made up of nearly 146 million bases, chromosome 8 encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and typically associated with a poor prognosis. Portions of chromosome 8 have been linked to schizophrenia and bipolar disorder. Trisomy 8, also known as Warkany syndrome 2, most often results in early miscarriage but is occasionally seen in a mosaic form in surviving patients who suffer to a varying degree from a number of symptoms including retarded mental and motor development, and certainMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno DPY19L4
Nombre del antígeno dpy-19-like 4 (C. elegans)
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 488
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 286148
Isotipo IgG
InmunoQuímica Yes
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR