Anti-CPXM1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Proveedor: Bioss
BS-8341R-HRP
BOSSBS-8341R-HRPEA 900 EUR
BOSSBS-8341R-HRP
Anti-CPXM1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticuerpos
CPXM (carboxypeptidase X, member 1) belongs to the peptidase M14 family. However, no carboxypeptidase activity has yet been detected. It may be involved in cell-cell interactions.Members of the M14 metallocarboxypeptidase protein family serve many diverse functions and are divided into three subfamilies based on structure, function and amino acid sequence similarity. Belonging to the N/E subfamily, CPXM (metallocarboxypeptidase CPX-1) is a 734 amino acid protein that contains a F5/8 type C domain and likely binds one zinc ion per subunit. Most members of the N/E subfamily contain severalMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno CPXM1
Nombre del antígeno Carboxypeptidase X (M14 Family), Member 1
Environmentally Preferable
Sinónimos de antígenos CPX1|CPXM
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado HRP (Horseradish Peroxidase)
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 56265
Isotipo IgG
Western blot Yes
InmunoQuímica Yes
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR