Anti-GCOM1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Proveedor: Bioss
BS-8403R-A555
BOSSBS-8403R-A555EA 893 EUR
BOSSBS-8403R-A555
Anti-GCOM1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticuerpos
Glutamate receptors mediate most excitatory neurotransmission in the brain and play an important role in neural plasticity, neural development and neurodegeneration. Ionotropic glutamate receptors are categorized into NMDA receptors and kainate/AMPA receptors, both of which contain glutamate-gated, cation-specific ion channels. Synaptic and extrasynaptic NMDA receptors have been shown to have opposite effects on neuronal survival, CREB function and gene regulation. Gcom1 (GRINL1A complex locus protein 1), also known as GUP (GRINL1A upstream protein) and Gcom (GRINL1A combined protein), is a 4More Product Information