Anti-RFTN2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Proveedor: Bioss
BS-8500R-FITC
BOSSBS-8500R-FITCEA 900 EUR
BOSSBS-8500R-FITC
Anti-RFTN2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticuerpos
Membrane microdomains known as lipid rafts are implicated in B-cell activation during B-cell receptor (BCR) signal initiation. Raftlin-2, also designated RFTN2 (raftlin family member 2) or raft-linking protein 2, is a 501 amino acid cell membrane protein that is essential for raft cell assembly and maintenance. A lipid anchor protein, Raftlin-2 belongs to the raftlin family and is encoded by a gene that maps to human chromosome 2q33.1 and mouse chromosome 1 C1.2. Human chromosome 2 is the second largest human chromosome, which consists of 237 million bases, encodes over 1,400 genes and makesMore Product Information