Anti-SDHC Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Proveedor: Bioss
BS-8530R-A488
BOSSBS-8530R-A488EA 893 EUR
BOSSBS-8530R-A488
Anti-SDHC Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticuerpos
Membrane-anchoring subunit of succinate dehydrogenase (SDH) that is involved in complex II of the mitochondrial electron transport chain and is responsible for transferring electrons from succinate to ubiquinone (coenzyme Q).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SDHC
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR