Anti-GRIN2B Rabbit Polyclonal Antibody (Cy3®)
Proveedor: Bioss
BS-8566R-CY3
BOSSBS-8566R-CY3EA 930 EUR
BOSSBS-8566R-CY3
Anti-GRIN2B Rabbit Polyclonal Antibody (Cy3®)
Anticuerpos
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA receptor channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of three different subunits: NR1 (GRIN1), NR2 (GRIN2A, GRIN2B, GRIN2C, or GRIN2D) and NR3 (GRIN3A or GRIN3B). The NR2 subunit acts as the agonist binding site for glutamate. This receptor is the predominant excitatory neurotransmitter receptor in theMore Product Information