Anti-SAMD3 Rabbit Polyclonal Antibody

Proveedor: Bioss
BS-9004R
BOSSBS-9004REA 725 EUR
BOSSBS-9004R
Anti-SAMD3 Rabbit Polyclonal Antibody
Anticuerpos
IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SAMD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR