Anti-EFHC2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Proveedor: Bioss
BS-9014R-A350
BOSSBS-9014R-A350EA 1350 EUR
BOSSBS-9014R-A350
Anti-EFHC2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticuerpos
EFHC2 contains 3 DM10 domains, followed by 3 putative EF-hand motifs. There are two named isoforms produced by alternative splicing. EFHC2 can be used as a quantitative trait locus for fear recognition in Turner syndrome.EF-HC2 is a 749 amino acid protein containing three DM10 domains and an EF-hand domain. EF-HC2 shares 41.6% homology with EF-HC1, and is widely expressed in peripheral tissues and central nervous system. The gene encoding EF-HC2 maps to human chromosome Xp11.3 and is critical for recognition of facial fear and harm avoidance. Turner syndrome, characterized by deficits inMore Product Information