Anti-SERPINB3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Proveedor: Bioss
BS-9214R-A350
BOSSBS-9214R-A350EA 893 EUR
BOSSBS-9214R-A350
Anti-SERPINB3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticuerpos
Metastasis of a primary tumor to a distant site is determined through signaling cascades that break down interactions between the cell and extracellular matrix proteins. Among the proteins mediating metastasis are serine prote-ases, such as neutrophil elastase.SCCA is transcribed by two nearly identical genes (SCCA1 and SCCA2), and is mainly produced as SCCA1. The human SCCA1 gene encodes a 390 amino acid protein that was originally isolated from a metastatic cervical squamous cell carcinoma.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SERPINB3
Clonality: Polyclonal
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR