Anti-RNF121 Rabbit Polyclonal Antibody (Cy5®)

Proveedor: Bioss
BS-9255R-CY5
BOSSBS-9255R-CY5EA 1350 EUR
BOSSBS-9255R-CY5
Anti-RNF121 Rabbit Polyclonal Antibody (Cy5®)
Anticuerpos
RNF121 protein contains a RING finger, a motif present in a variety of functionally distinct proteins and known to be involved in protein-protein and protein-DNA interactions. Multiple alternatively spliced transcript variants encoding distinct isoforms have been reported. Three of them are supported by at least two independent transcripts or ESTs, the full length natures of others are not clear.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RNF121
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno RNF121
Environmentally Preferable
Nombre del antígeno Ring Finger Protein 121
Sinónimos de antígenos 4930544L10Rik|FLJ11099|RN121_HUMAN.
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Cy5®
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 55298
Isotipo IgG
InmunoQuímica Yes
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR