Anti-SLC25A37 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Proveedor: Bioss
BS-9523R-A350
BOSSBS-9523R-A350EA 893 EUR
BOSSBS-9523R-A350
Anti-SLC25A37 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticuerpos
Mitoferrin1, SLC25A37, belongs to the mitochondrial carrier family and contains 3 Solcar repeats. It is a mitochondrial iron transporter that specifically mediates iron uptake in developing erythroid cells and plays an essential role in heme biosynthesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC25A37
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

ESPECIFICACIONES

Simbolo antígeno SLC25A37
Environmentally Preferable
Nombre del antígeno Solute Carrier Family 25 Member A37
Sinónimos de antígenos MFRN1|MSCP|PRO1584|PRO2217|HT015|PRO1278|MSC|MFRN
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 350
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 51312
Isotipo IgG
InmunoQuímica Yes
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR