Anti-PARP16 Rabbit Polyclonal Antibody (Cy7®)
Proveedor: Bioss
BS-9623R-CY7
BOSSBS-9623R-CY7EA 900 EUR
BOSSBS-9623R-CY7
Anti-PARP16 Rabbit Polyclonal Antibody (Cy7®)
Anticuerpos
Poly(ADP-ribosylation) is a method of DNA damage-dependent posttranslational modification that helps to rescue injured proliferating cells from cell death. The PARP (poly(ADP-ribose) polymerase) proteins comprise a superfamily of enzymes that functionally modify histones and other nuclear proteins, thereby preventing cell death. PARPs use NAD+ as a substrate to catalytically transfer ADP-ribose residues onto protein acceptors; a process that, when repeated multiple times, leads to the formation of poly(ADPribose) chains on the protein. The presence of these chains alters the function of theMore Product Information