Anti-FAM76A Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Proveedor: Bioss
BS-9658R-A555
BOSSBS-9658R-A555EA 893 EUR
BOSSBS-9658R-A555
Anti-FAM76A Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticuerpos
Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. TheMore Product Information
Order Now

ESPECIFICACIONES

Simbolo antígeno FAM76A
Nombre del antígeno Family With Sequence Similarity 76, Member A
Environmentally Preferable
Tipo de anticuerpo Primario
Clonalidad Polyclonal
Conjugado Alexa Fluor® 555
Reactividad Human,Rat,Mouse
Huésped Rabbit
ID del gen 199870
Isotipo IgG
InmunoQuímica Yes
Forma liquid
Purification Purified by Protein A
Concentración 1 μg/μl
Adsorción cruzada No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Temperatura de almacenamiento Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR