Anti-ATP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Proveedor: Bioss
BS-9885R-A680
BOSSBS-9885R-A680EA 893 EUR
BOSSBS-9885R-A680
Anti-ATP1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anticuerpos
TMEM103, also known as C3orf75, is a 266 amino acid protein encoded by a gene mapping to human chromosome 3. Chromosome 3 is made up of about 214 million bases encoding over 1100 genes. Notably, there is a chemokine receptor gene cluster and a variety of human cancer related loci on chromosome 3. Particular regions of the chromosome 3 short arm are deleted in many types of cancer cells. Key tumour suppressing genes on chromosome 3 encode apoptosis mediator RASSF1, cell migration regulator HYAL1 and angiogenesis suppressor SEMA3B. Marfan Syndrome, porphyria, von Hippel-Lindau syndrome,More Product Information