EMC1 Mouse Monoclonal Antibody [clone: 2F3] (ALEXA FLUOR® 680)

Proveedor: Bioss
BSM-2279M-A680
BOSSBSM-2279M-A680EA 1090 EUR
BOSSBSM-2279M-A680
EMC1 Mouse Monoclonal Antibody [clone: 2F3] (ALEXA FLUOR® 680)
Anticuerpos
This gene encodes a single-pass type I transmembrane protein, which is a subunit of the endoplasmic reticulum membrane protein complex (EMC). Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Type: Primary
Antigen: ER membrane protein complex subunit 1
Clonality: Monoclonal
Clone: 2F3
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/993
Host: Mouse
Isotype: IgG
Reactivity: Human; Rat; Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR