BradyPrinter i5300 Industrial Label Printers
Proveedor: Brady
151274 151275 151283 151284 322900 322891 322894 322897 322899 322890 322893
BRDY151274EA 2280 EUR
BRDY151274 BRDY151275 BRDY151283 BRDY151284 BRDY322900 BRDY322891 BRDY322894 BRDY322897 BRDY322899 BRDY322890 BRDY322893
BradyPrinter i5300 Industrial Label Printers
Impresoras Label Printers
Zero calibration, zero wasted labels, with higher-volume duty cycle and ultra-precise printing. Prints on a very wide range of B30/B33 materials with exceptional accuracy on labels as small as 5 mm. For ultimate high-mix printing with 20 second no-adjustment material changeovers, built-in auto-cutting and optional save-to-printer job lists for a quick way to print commonly used labels and overwrite-protected labels away from the PC. When focused on efficiency, every process counts - let simplicity drive productivity.
  • No wasted labels - prints on the first one
  • i5300 - 20-second supply