Brady® FR22 RFID Fixed Reader
Proveedor: Brady
311937
BRDY311937EA 2670 EUR
BRDY311937
Brady® FR22 RFID Fixed Reader
Códigos de barras
The BRADY FR22 is the core device of a new approach for RFID fixed systems with wide communication options and a modular design. The FR22 is a critical building block for various track and trace and intelligent manufacturing solutions. It enables different use cases for different segments depending on which modules are attached to it.
  • Fast and reliable edge computing with support for custom embedded applications
  • New UHF RFID engine based on the very latest technology
  • Brady extension connector for plug and play accessories like Brady MUX16
  • Connect external
More Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable
Temperatura de funcionamiento −20...+55 °C
Alimentación 100 - 240 V at 50 - 60 Hz
W×D×H 203,2×78,49×17,55 mm