Magicard™ Colour Film for Pronto 100, 100 Shot
Proveedor: Brady
321975
BRDY321975EA 52.5 EUR
BRDY321975
Magicard™ Colour Film for Pronto 100, 100 Shot
Cintas de impresora
The Magicard™ colour ribbon is perfect for full colour prints with a Pronto 100 printer.
  • Overlay panel for extra print protections
  • Print 100 cards in one go
This high-quality, YMCKO ribbon guarantees to print 100 cards on the Pronto 100 printer.
Información de suministro: Includes 1 dye film and 1 cleaning roller.
New Product