Barcode Scanner, V4500
Proveedor: Brady
177233 322619 177231 322621 322623
BRDY177233EA 805 EUR
BRDY177233 BRDY322619 BRDY177231 BRDY322621 BRDY322623
Barcode Scanner, V4500
Códigos de barras
A rugged design offering seamless experience. Digitise data capture and automate your workflows with ease and accuracy with the V4500 Barcode Scanner.
  • High-speed, omnidirectional barcode scanning made simple
  • Ideal for inventory management, kit verification, scan and print, and packout applications
  • Proprietary cortex decoder technology allows scanner to easily read tiny barcodes as well as shiny and hard-to-read surfaces
  • Automated keyboard device and dynamic user interface lets you go from setup to first scan in seconds
  • Data parsing feature lets you sort
More Product Information
New Product
Order Now
ESPECIFICACIONES
Environmentally Preferable
Interfaz RS232, USB 2.0 (generic HID, HID keyboard, virtual com port) RS232, USB 2.0 (generic HID, HID keyboard, virtual com port)
Tipo de sensor CMOS 1.2 Megapixel CMOS 1.2 Megapixel
Alimentación 100 - 240 V, 50 - 60 Hz 100 - 240 V, 50 - 60 Hz
W×D×H 86×160×198 mm 152×113×75 mm
Peso 0,53 kg 0,53 kg