Insulation plugs for industrial applications
Proveedor: Brady
812713 812711 812712 850958
BRDY812713EA 30.1 EUR
BRDY812713 BRDY812711 BRDY812712 BRDY850958
Insulation plugs for industrial applications
Cierres de seguridad
These insulation plugs protect people working on current circuits against unauthorised use and inadvertent switching on of power. The insulation plug is placed into a fuse base by means of a special screwdriver (sold separately) instead of fuse link and screw cap.
Información para pedidos: Available individually or as a set inclunding insulation plugs DII and DIII (3 each) and a screwdriver.