Graduated pipettes, BLAUBRAND®, type 1, partial delivery (one batch)
Proveedor: Brand
2.27722 2.27725 2.27727 2.27729 2.27731 2.27721
BRND2.27722EA 95.1 EUR
BRND2.27722 BRND2.27725 BRND2.27727 BRND2.27729 BRND2.27731 BRND2.27721
Graduated pipettes, BLAUBRAND®, type 1, partial delivery (one batch)
Pipetas Pipetas graduadas
AR-Glas®.
  • Calibrated to deliver (TD, Ex;)
  • Only marked pipettes (*) are suitable for cotton plugs
  • Incl. one batch certificate
  • Marks and inscriptions in high contrast blue enamel
Length: 360 mm ±10 mm except for 25 ml pipettes: 450 mm ±10 mm.
Certificaciones: DIN EN ISO 835. DE-M marking.
Información de suministro: All supplied pipettes are from the same batch.