Volumetric flasks, class A, trapezoidal shape, BLAUBRAND®
Proveedor: Brand
2.36408 2.36403 2.36404 2.36406 2.36407 2.36402 2.36401
BRND2.36408EA 61.8 EUR
BRND2.36408 BRND2.36403 BRND2.36404 BRND2.36406 BRND2.36407 BRND2.36402 BRND2.36401
Volumetric flasks, class A, trapezoidal shape, BLAUBRAND®
Matraces Matraces aforados
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • DE-M marking
  • Total order from one batch, batch certificate included
*Wide neck.
Certificaciones: DIN EN ISO 1042