Volumetric flasks, class A, blue markings, BLAUBRAND®
Proveedor: Brand
2.37253 2.37291 2.37290 2.37236 2.37254 2.37255 2.37245 2.37234 2.37252 2.37251 2.37250 2.37249 2.37248 2.37247 2.37246 2.37243 2.37238
612-3617EA 69.4 EUR
612-3617 BRND2.37291 BRND2.37290 BRND2.37236 BRND2.37254 BRND2.37255 BRND2.37245 BRND2.37234 BRND2.37252 BRND2.37251 BRND2.37250 BRND2.37249 BRND2.37248 BRND2.37247 BRND2.37246 BRND2.37243 BRND2.37238
Volumetric flasks, class A, blue markings, BLAUBRAND®
Matraces Matraces aforados
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in high contrast blue enamel
  • marcaje DE-M
  • Total order from one batch, supplied with batch certificate
* In addition to DIN.
Certificaciones: DIN EN ISO 1042.