Cubetas de tinción, Schiefferdecker
Proveedor: Brand
472500
BRND472500EA 339 EUR
BRND472500
Cubetas de tinción, Schiefferdecker
Tarros Cubetas de tinción
Jars , made of soda-lime glass, with lid.
These staining trough hold up to 10 slides (76×26 mm), or 10 pairs of slides placed adjacently back-to-back.
Jars made of heavy-walled soda lime glass with a relatively high coefficient of expansion, and it should not exposed to rapid temperature changes, to avoid cracks caused by tensions.