Measuring cylinders, tall form, class A, borosilicate glass, USP, BLAUBRAND®
Proveedor: Brand
32805 32808 32828 32820 32838 32848 32854 32862 32864 USP32864 USP32862 USP32854 USP32848 USP32838 USP32820 USP32808 USP32805
612-4834EA 33.3 EUR
612-4834 612-4835 612-4837 612-4836 612-4838 612-4839 612-4840 612-4841 612-4842 BRNDUSP32864 BRNDUSP32862 BRNDUSP32854 BRNDUSP32848 BRNDUSP32838 BRNDUSP32820 BRNDUSP32808 612-5747
Measuring cylinders, tall form, class A, borosilicate glass, USP, BLAUBRAND®
Probetas Probetas graduadas
Borosilicate glass 3,3, tall form, class A, conformity certified.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, with spout
  • Ring marks at major graduations, marks and inscriptions in high contrast blue enamel
DIN EN ISO 4788
Certificaciones: DIN EN ISO 4788