Anti-DHODH Rabbit Polyclonal Antibody

Proveedor: Boster Bio
PB10056
BSBTPB10056EA 552 EUR
BSBTPB10056
Anti-DHODH Rabbit Polyclonal Antibody
Anticuerpos
Rabbit IgG polyclonal antibody for Dihydroorotate dehydrogenase (quinone), mitochondrial(DHODH) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Type: Primary
Antigen: DHODH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR