Anti-Peroxisome proliferator-activated receptor gamma Chicken Polyclonal Antibody
Proveedor: Biosensis
C-1533-100
BSENC-1533-100EA 321 EUR
BSENC-1533-100
Anti-Peroxisome proliferator-activated receptor gamma Chicken Polyclonal Antibody
Anticuerpos
Peroxisome proliferator-activated receptor gamma (PPARG) is a nuclear hormone receptor that binds peroxisome proliferators such as hypolipidemic drugs and fatty acids. Once activated by a ligand, the receptor binds to a promoter element in the gene for acyl-CoA oxidase and activates its transcription (Ref: SWISSPROT).
Antibody detects PPARG by western blot in human cell nuclear extracts. It is recommended to isolate nuclei from stimulated cultures for best results.
Application Information:
WB. Suggested dilution at 1:2,000 to 1:5,000. Biosensis recommends that the optimal working dilutionMore Product Information