Anti-Atrial Natriuretic Peptide Rabbit Polyclonal Antibody
Proveedor: Biosensis
R-1500-50
BSENR-1500-50EA 321 EUR
BSENR-1500-50
Anti-Atrial Natriuretic Peptide Rabbit Polyclonal Antibody
Anticuerpos
The hormone Atrial natriuretic peptide (ANP) is produced by cardiomyocites and is secreted into the bloodstream. ANP acts on convoluted tubules in kidney reducing sodium absorption which results in reducing the blood pressure.
The antibody recognizes ANP by native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by theMore Product Information