Anti-Neurotensin Rabbit Polyclonal Antibody
Proveedor: Biosensis
R-1503-50
BSENR-1503-50EA 321 EUR
BSENR-1503-50
Anti-Neurotensin Rabbit Polyclonal Antibody
Anticuerpos
Neurotensin is a neuropeptide which regulates secretion of luteinizing hormone and prolactin. Involved in dopamine neurotransmission.
Antibody recognizes Neurotensin by native applications. A dilution of 5-10 µg/ml is recommended for immunohistochemistry using formalin fixed and paraffin embedded tissues and for 4% paraformaldehyde fixed frozen tissues. A dilution of 5-15 µg/ml is recommended for immunofluorescence. . Biosensis recommends optimal dilutions/concentrations should be determined by the end user. Cross reactivity: Human; mouse; rat. Highly conserved and expected to interactMore Product Information