Sheep Monoclonal IgG

Proveedor: Biosensis
S-1754-500
BSENS-1754-500EA 112 EUR
BSENS-1754-500
Sheep Monoclonal IgG
Anticuerpos
Sheep serum from non-immunised animals was purified by protein G affinity chromatography. This sheep IgG fraction can be used as negative control antibody for various immunological techniques where sheep primary antibodies are used.

Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR