Parallel synthesis, Syncore® polyvap
Proveedor: BUCHI
1P3S23000 1P3002000 1P3003000 1P3004000 1P3005000 1P3006000 1P3007000 1P3008000 1P3C00000 1P3C01000 1P3C02000 1P3C03000 1P3C04000 1P3C05000 1P3C06000 1P3C10000 1P3C11000 1P3C12000 1P3C13000 1P3C14000 1P3C15000 1P3C16000 1P3C17000 1P3C20000 1P3C21000 1P3C22000 1P3C23000 1P3C24000 1P3C25000 1P3C26000 1P3C27000 1P3C28000 1P3S00000 1P3S01000 1P3S02000 1P3S03000 1P3S04000 1P3S05000 1P3S06000 1P3S07000 1P3S08000 1P3S10000 1P3S11000 1P3S12000 1P3S13000 1P3S14000 1P3S15000 1P3S16000 1P3S17000 1P3S20000 1P3S21000 1P3S22000 1P3S24000 1P3S25000 1P3S26000 1P3S27000 1P3S28000 1P3000000 1P3001000
BUCH1P3S23000EA 0 EUR
BUCH1P3S23000 BUCH1P3002000 BUCH1P3003000 BUCH1P3004000 BUCH1P3005000 BUCH1P3006000 BUCH1P3007000 BUCH1P3008000 BUCH1P3C00000 BUCH1P3C01000 BUCH1P3C02000 BUCH1P3C03000 BUCH1P3C04000 BUCH1P3C05000 BUCH1P3C06000 BUCH1P3C10000 BUCH1P3C11000 BUCH1P3C12000 BUCH1P3C13000 BUCH1P3C14000 BUCH1P3C15000 BUCH1P3C16000 BUCH1P3C17000 BUCH1P3C20000 BUCH1P3C21000 BUCH1P3C22000 BUCH1P3C23000 BUCH1P3C24000 BUCH1P3C25000 BUCH1P3C26000 BUCH1P3C27000 BUCH1P3C28000 BUCH1P3S00000 BUCH1P3S01000 BUCH1P3S02000 BUCH1P3S03000 BUCH1P3S04000 BUCH1P3S05000 BUCH1P3S06000 BUCH1P3S07000 BUCH1P3S08000 BUCH1P3S10000 BUCH1P3S11000 BUCH1P3S12000 BUCH1P3S13000 BUCH1P3S14000 BUCH1P3S15000 BUCH1P3S16000 BUCH1P3S17000 BUCH1P3S20000 BUCH1P3S21000 BUCH1P3S22000 BUCH1P3S24000 BUCH1P3S25000 BUCH1P3S26000 BUCH1P3S27000 BUCH1P3S28000 BUCH1P3000000 BUCH1P3001000
Parallel synthesis, Syncore® polyvap
Evaporadores Sistemas para síntesis paralela
Highly modular parallel evaporator with interchangeable racks for 4 to 96 samples, covering working volumes of 0,5 to 500 ml. Due to a number of optional modular asseccories the Polyvap is instantly convertable into the Syncore® Reactor performing synthetic procedures or the Syncore® Analyst for concentration to pre-defined residual volumes.
  • Boiling retardation is prevented during evaporation
  • Possibility of cross-contamination is eliminated by a specially designed vacuum cover
  • Relevant parts of the Polyvap are chemically resistant and free of contaminants,
More Product Information
Order Now
ESPECIFICACIONES
Modelo Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap Polyvap
Environmentally Preferable
Potencia de calefacción 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W 1500 W
Intervalo de temperaturas Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C Ambient…150 °C
An × Pr 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm 480×487 mm