Flow meters
Proveedor: BURKLE
5700-0141 5700-0121 5700-0111
BURK5700-0141EA 1550 EUR
BURK5700-0141 BURK5700-0121 BURK5700-0111
Flow meters
Medidores de flujo
These flow meters are compact and fully electronic for electric drum and container pumps, also used for low viscosity media.
  • Measurement by the turbine wheel principle
  • Can be calibrated depending on the medium
  • Different versions for many different applications
  • Automatic switch on when flow is recognized
  • Four resettable part ranges and one fixed total range