Anti-Cox-1 Rabbit Polyclonal Antibody [clone: S585]

Proveedor: Bioworld Technology
BS1075
BWRLBS1075EA 262 EUR
BWRLBS1075
Anti-Cox-1 Rabbit Polyclonal Antibody [clone: S585]
Anticuerpos
Synthetic peptide, corresponding to amino acids 555-600 of Human Cox-1.

Cox1 (S585) pAb detects endogenous levels of Cox1 protein.

Type: Primary
Antigen: Cox-1
Clonality: Polyclonal
Clone: S585
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR