Anti-Tubulin beta Mouse Monoclonal Antibody [clone: 4D9 mAb-HRP]

Proveedor: Bioworld Technology
BS1482MH
BWRLBS1482MHEA 246 EUR
BWRLBS1482MH
Anti-Tubulin beta Mouse Monoclonal Antibody [clone: 4D9 mAb-HRP]
Anticuerpos
Synthetic peptide, corresponding to amino acids 402-455 of Human Tubulin beta.

Tubulin ? (4D9) mAb-HRP detects endogenous levels of ?-Tubulin protein.

Type: Primary
Antigen: Tubulin beta
Clonality: Monoclonal
Clone: 4D9 mAb-HRP
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR