Anti-TRH-R1 Rabbit Polyclonal Antibody [clone: W234]

Proveedor: Bioworld Technology
BS3241
BWRLBS3241EA 262 EUR
BWRLBS3241
Anti-TRH-R1 Rabbit Polyclonal Antibody [clone: W234]
Anticuerpos
Synthetic peptide, corresponding to amino acids 250-300 of Human TRH-R1.

TRH-R1 (W234) pAb detects endogenous levels of TRH-R1 protein.

Type: Primary
Antigen: TRH-R1
Clonality: Polyclonal
Clone: W234
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR