Anti-MRP-S34 Rabbit Polyclonal Antibody [clone: K214]

Proveedor: Bioworld Technology
BS3327
BWRLBS3327EA 262 EUR
BWRLBS3327
Anti-MRP-S34 Rabbit Polyclonal Antibody [clone: K214]
Anticuerpos
Synthetic peptide, corresponding to amino acids 180-226 of Human MRP-S34.

MRP-S34 (K214) pAb detects endogenous levels of MRP-S34 protein.

Type: Primary
Antigen: MRP-S34
Clonality: Polyclonal
Clone: K214
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR